Assay Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014042.1 | NP_001014064.1 | 1 - 2 | 273 | 122 | |
| XM_006233150.2 | XP_006233212.1 | 1 - 2 | 639 | 122 | ||
| GenBank mRNA | BC087113.1 | - | 1 - 2 | 273 | 7193587 | 122 |
More Information
Updated Mapping Information
Set Membership: |
|

Mybphl
Genomic Map