Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033079.1 | NP_001028251.1 | 10 - 11 | 2126 | 63 | |
| GenBank mRNA | L06434.2 | - | 10 - 11 | 2126 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a17
Genomic Map