Assay Details
| Gene Symbol: Amy1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010970.1 | NP_001010970.1 | 1 - 2 | 47 | 88 | |
| GenBank mRNA | AB057450.1 | - | 1 - 2 | 47 | 88 | |
| FQ219213.1 | - | 1 - 2 | 376 | 88 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB057450.1,NM_001010970.1 transcripts.
Set Membership: |
|

Amy1a
Genomic Map