Assay Details
| Gene Symbol: Rtcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004227.1 | NP_001004227.1 | 3 - 4 | 303 | 65 | |
| XM_008761456.1 | XP_008759678.1 | 2 - 3 | 259 | 65 | ||
| GenBank mRNA | BC079374.1 | - | 3 - 4 | 303 | 7110129 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Rtcd1
Genomic Map