Assay Details
| Gene Symbol: Rtcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004227.1 | NP_001004227.1 | 6 - 7 | 633 | 102 | |
| XM_008761456.1 | XP_008759678.1 | 5 - 6 | 589 | 102 | ||
| GenBank mRNA | BC079374.1 | - | 6 - 7 | 633 | 7110129 | 102 |
More Information
Updated Mapping Information
Set Membership: |
|

Rtcd1
Genomic Map