Assay Details
| Gene Symbol: Lppr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001508.2 | NP_001001508.2 | 1 - 2 | 813 | 68 | |
| GenBank mRNA | AF541280.1 | - | 1 - 2 | 814 | 68 | |
| AY266268.1 | - | 1 - 2 | 814 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lppr4
Genomic Map