Assay Details
| Gene Symbol: Gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233424.2 | XP_006233486.1 | 9 - 10 | 1798 | 68 | |
| XM_227762.8 | XP_227762.6 | 9 - 10 | 1845 | 68 | ||
| GenBank mRNA | FQ225190.1 | - | 9 - 10 | 1621 | 68 | |
More Information
Set Membership: |
|

Gbp4
Genomic Map