Assay Details
| Gene Symbol: Gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233424.2 | XP_006233486.1 | 10 - 11 | 1989 | 143 | |
| XM_227762.8 | XP_227762.6 | 10 - 11 | 2036 | 143 | ||
| GenBank mRNA | FQ225190.1 | - | 10 - 11 | 1812 | 143 | |
More Information
Set Membership: |
|

Gbp4
Genomic Map