Assay Details
| Gene Symbol: Gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233424.2 | XP_006233486.1 | 5 - 6 | 1004 | 60 | |
| XM_227762.8 | XP_227762.6 | 5 - 6 | 1006 | 60 | ||
| GenBank mRNA | FQ225190.1 | - | 5 - 6 | 782 | 60 | |
| Gene Symbol: LOC100911495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761539.1 | XP_008759761.1 | 998 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gbp4
Genomic Map