Assay Details
| Gene Symbol: Gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233424.2 | XP_006233486.1 | 7 - 8 | 1481 | 100 | |
| XM_227762.8 | XP_227762.6 | 7 - 8 | 1528 | 100 | ||
| GenBank mRNA | FQ225190.1 | - | 7 - 8 | 1304 | 100 | |
More Information
Set Membership: |
|

Gbp4
Genomic Map