Assay Details
| Gene Symbol: Clca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013202.2 | NP_001013220.1 | 4 - 5 | 573 | 76 | |
| GenBank mRNA | AB119249.1 | - | 4 - 5 | 574 | 76 | |
| AB212889.1 | - | 4 - 5 | 574 | 76 | ||
| AB306511.1 | - | 4 - 5 | 574 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clca2
Genomic Map