Assay Details
| Gene Symbol: Ccbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015037.1 | NP_001015037.1 | 183 | 103 | ||
| GenBank mRNA | AY955395.1 | - | 183 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl2
Genomic Map