Assay Details
| Gene Symbol: Ptgfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233486.2 | XP_006233548.1 | 1 - 2 | 112 | 75 | |
| GenBank mRNA | X83856.1 | - | 1 - 2 | 83 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the X83856.1, XM_006233486.2 transcripts.
Set Membership: |
|

Ptgfr
Genomic Map