Assay Details
| Gene Symbol: Gnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108950.2 | NP_001102420.2 | 3 - 4 | 243 | 69 | |
| XM_006233171.1 | XP_006233233.1 | 3 - 4 | 276 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnat2
Genomic Map