Assay Details
| Gene Symbol: LOC100911379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233104.2 | XP_006233166.1 | 5 - 6 | 1174 | 67 | |
| XM_008761437.1 | XP_008759659.1 | 730 | 67 | |||
| Gene Symbol: LOC100363228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233106.2 | XP_006233168.1 | 5 - 6 | 514 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911379
Genomic Map