Assay Details
Gene Symbol: NEWGENE_1305281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233223.2 | XP_006233285.1 | 6 - 7 | 966 | 72 | |
XM_006233224.2 | XP_006233286.1 | 6 - 7 | 1081 | 72 | ||
XM_008761473.1 | XP_008759695.1 | 6 - 7 | 837 | 72 |
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106467.3 | NP_001099937.1 | 834 | 72 | ||
GenBank mRNA | BC160912.1 | - | 823 | 7456962 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
