Assay Details
| Gene Symbol: Clca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201419.1 | NP_958822.1 | 3 - 4 | 462 | 144 | |
| GenBank mRNA | AY333961.1 | - | 3 - 4 | 462 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clca4
Genomic Map