Assay Details
| Gene Symbol: Clca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201419.1 | NP_958822.1 | 5 - 6 | 745 | 123 | |
| GenBank mRNA | AY333961.1 | - | 5 - 6 | 745 | 123 | |
More Information
Set Membership: |
|

Clca4
Genomic Map