Assay Details
| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106475.1 | NP_001099945.1 | 1 - 2 | 155 | 78 | |
| XM_006233371.2 | XP_006233433.1 | 2 - 3 | 237 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adh6a
Genomic Map