Assay Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044303.2 | NP_001037768.2 | 12 - 13 | 2147 | 68 | |
| XM_008761422.1 | XP_008759644.1 | 12 - 13 | 3328 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P022290 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P078210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078210 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022290 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078210 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P022290 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P022290 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P078210 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52625 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39395 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10818420 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818424 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818421 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818423 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818418 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818422 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818413 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818419 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818415 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10818417 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10818412 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200013884.rn.1 | TC0200002004.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200013883.rn.1 | TC0200002004.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10818412 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21210142 | - | - |
| ABI Rat Genome Survey Microarray | 21287535 | - | - |

Aknad1
Genomic Map