Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133624.2 | NP_598308.1 | 1 - 2 | 152 | 73 | |
| GenBank mRNA | BC086991.1 | - | 1 - 2 | 114 | 7191761 | 73 |
| M80367.1 | - | 1 - 2 | 153 | 73 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC086991.1,M80367.1,NM_133624.2 transcripts.
Set Membership: |
|

Gbp2
Genomic Map