Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133624.2 | NP_598308.1 | 3 - 4 | 497 | 67 | |
| GenBank mRNA | BC086991.1 | - | 3 - 4 | 459 | 7191761 | 67 |
| M80367.1 | - | 3 - 4 | 498 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gbp2
Genomic Map