Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133624.2 | NP_598308.1 | 7 - 8 | 1325 | 97 | |
| GenBank mRNA | BC086991.1 | - | 7 - 8 | 1287 | 7191761 | 97 |
| M80367.1 | - | 7 - 8 | 1326 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gbp2
Genomic Map