Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133624.2 | NP_598308.1 | 9 - 10 | 1643 | 61 | |
| GenBank mRNA | BC086991.1 | - | 9 - 10 | 1605 | 7191761 | 61 |
| FQ223354.1 | - | 2 - 3 | 288 | 61 | ||
| M80367.1 | - | 9 - 10 | 1644 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gbp2
Genomic Map