Assay Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021664.1 | NP_067696.1 | 3 - 4 | 528 | 78 | |
| XM_006233464.2 | XP_006233526.1 | 4 - 5 | 605 | 78 | ||
| XM_006233465.2 | XP_006233527.1 | 4 - 5 | 632 | 78 | ||
| XM_006233466.2 | XP_006233528.1 | 4 - 5 | 635 | 78 | ||
| XM_006233468.2 | XP_006233530.1 | 4 - 5 | 1016 | 78 | ||
| XM_008761521.1 | XP_008759743.1 | 4 - 5 | 738 | 78 | ||
| GenBank mRNA | AF178974.1 | - | 3 - 4 | 528 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnase2b
Genomic Map