Assay Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF063102.1 | - | 12 - 13 | 2364 | 72 | |
| AF081148.1 | - | 13 - 14 | 2810 | 72 | ||
| AF081149.1 | - | 13 - 14 | 2810 | 72 | ||
| AF081150.1 | - | 13 - 14 | 2810 | 72 | ||
| AF081151.1 | - | 13 - 14 | 2810 | 72 | ||
| AF081152.1 | - | 13 - 14 | 2810 | 72 | ||
| AF081153.1 | - | 13 - 14 | 2810 | 72 | ||
More Information
Set Membership: |
|

Lphn2
Genomic Map