Assay Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF081151.1 | - | 19 - 20 | 3623 | 124 | |
| AF081152.1 | - | 19 - 20 | 3623 | 124 | ||
| AF081153.1 | - | 19 - 20 | 3623 | 124 | ||
More Information
Set Membership: |
|

Lphn2
Genomic Map