Assay Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF063102.1 | - | 22 - 23 | 3599 | 81 | |
| AF081150.1 | - | 21 - 22 | 3973 | 81 | ||
| AF081153.1 | - | 22 - 23 | 4018 | 81 | ||
More Information
Set Membership: |
|

Lphn2
Genomic Map