Assay Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF081148.1 | - | 18 - 19 | 3579 | 117 | |
| AF081149.1 | - | 18 - 19 | 3579 | 117 | ||
| AF081150.1 | - | 18 - 19 | 3579 | 117 | ||
More Information
Set Membership: |
|

Lphn2
Genomic Map