Assay Details
| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039609.1 | NP_001034698.1 | 515 | 58 | ||
| GenBank mRNA | AB250295.1 | - | 515 | 58 | ||
| AB250296.1 | - | 515 | 58 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
membrane budding
autophagy
nervous system development
membrane invagination
vesicle organization
vesicle transport along actin filament
cell differentiation
cilium assembly
positive regulation of filopodium assembly
clathrin-mediated endocytosis
membrane tubulation
lipid binding
endocytosis
membrane budding
autophagy
nervous system development
membrane invagination
vesicle organization
vesicle transport along actin filament
cell differentiation
cilium assembly
positive regulation of filopodium assembly
clathrin-mediated endocytosis
membrane tubulation
lipid binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151547 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P081987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132892 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151547 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081987 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151547 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081987 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P081987 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151547 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P168564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68667 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18243 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70678 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75976 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84432 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10826511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826512 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10826502 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200032018.rn.1 | TC0200004599.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10826502 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355243 | - | - |

Fnbp1l
Genomic Map