Assay Details
| Gene Symbol: Obp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173148.1 | NP_775171.1 | 550 | 82 | ||
| GenBank mRNA | M76734.1 | - | 550 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Obp2b
Genomic Map