Assay Details
| Gene Symbol: Sgol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766787.1 | XP_008765009.1 | 8 - 9 | 1714 | 188 | |
| XM_008766788.1 | XP_008765010.1 | 8 - 9 | 1683 | 188 | ||
| XM_008766789.1 | XP_008765011.1 | 8 - 9 | 1711 | 188 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sgol1
Genomic Map