Assay Details
| Gene Symbol: LOC100911190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771241.1 | XP_008769463.1 | 4 - 5 | 1666 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008771241.1 transcripts.
Set Membership: |
|

LOC100911190
Genomic Map