Assay Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019319.1 | NP_062192.1 | 1 - 2 | 548 | 108 | |
| XM_008772166.1 | XP_008770388.1 | 2 - 3 | 1497 | 108 | ||
| XM_008772167.1 | XP_008770389.1 | 2 - 3 | 1446 | 108 | ||
| GenBank mRNA | Z21935.1 | - | 1 - 2 | 470 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk4
Genomic Map