Assay Details
Gene Symbol: Unc93a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001061886.4 | XP_001061886.2 | 3 - 4 | 706 | 94 | |
XM_008758782.1 | XP_008757004.1 | 3 - 4 | 542 | 94 | ||
XM_008758783.1 | XP_008757005.1 | 4 - 5 | 1022 | 94 | ||
XM_008758784.1 | XP_008757006.1 | 5 - 6 | 683 | 94 | ||
XM_008758785.1 | XP_008757007.1 | 3 - 4 | 1454 | 94 | ||
XM_008758786.1 | XP_008757008.1 | 6 - 7 | 972 | 94 | ||
XM_008758787.1 | XP_008757009.1 | 4 - 5 | 1305 | 94 | ||
XM_008758788.1 | XP_008757010.1 | 3 - 4 | 919 | 94 | ||
XM_008758789.1 | XP_008757011.1 | 3 - 4 | 1693 | 94 | ||
XM_008758790.1 | XP_008757012.1 | 4 - 5 | 609 | 94 | ||
XM_008758791.1 | XP_008757013.1 | 3 - 4 | 248 | 94 | ||
XM_008758792.1 | XP_008757014.1 | 4 - 5 | 773 | 94 | ||
XM_008758793.1 | XP_008757015.1 | 3 - 4 | 1683 | 94 | ||
XM_008758794.1 | XP_008757016.1 | 3 - 4 | 350 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
