Assay Details
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034912.1 | NP_001030084.1 | 915 | 82 | ||
GenBank mRNA | AY310142.1 | - | 304 | 82 | ||
AY321329.1 | - | 299 | 82 | |||
BC089887.1 | - | 915 | 7371502 | 82 |
Gene Symbol: LOC103689968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232083.2 | XP_006232145.1 | 6 - 7 | 592 | 82 | |
XM_006232084.2 | XP_006232146.1 | 5 - 6 | 958 | 82 | ||
XM_008760821.1 | XP_008759043.1 | 7 - 8 | 1009 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
