Assay Details
| Gene Symbol: Rsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232102.2 | XP_006232164.1 | 3 - 4 | 522 | 74 | |
| XM_006232103.2 | XP_006232165.1 | 3 - 4 | 791 | 74 | ||
| XM_006232104.2 | XP_006232166.1 | 3 - 4 | 246 | 74 | ||
| XM_006232105.2 | XP_006232167.1 | 3 - 4 | 546 | 74 | ||
| XM_006232106.2 | XP_006232168.1 | 3 - 4 | 254 | 74 | ||
| XM_008760847.1 | XP_008759069.1 | 3 - 4 | 834 | 74 | ||
More Information
Set Membership: |
|

Rsl1
Genomic Map