Assay Details
| Gene Symbol: Epb41l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227709.1 | XP_006227771.1 | 20 - 21 | 3229 | 57 | |
| XM_008758646.1 | XP_008756868.1 | 20 - 21 | 3411 | 57 | ||
| XM_008758647.1 | XP_008756869.1 | 20 - 21 | 3527 | 57 | ||
| XM_008758648.1 | XP_008756870.1 | 20 - 21 | 3132 | 57 | ||
| XM_008758649.1 | XP_008756871.1 | 21 - 22 | 3367 | 57 | ||
| XM_008758650.1 | XP_008756872.1 | 20 - 21 | 6214 | 57 | ||
| GenBank mRNA | FQ212032.1 | - | 16 - 17 | 2593 | 57 | |
More Information
Set Membership: |
|

Epb41l2
Genomic Map