Assay Details
| Gene Symbol: Dpy19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242722.2 | XP_006242784.1 | 13 - 14 | 1563 | 77 | |
| XM_235972.8 | XP_235972.6 | 13 - 14 | 1563 | 77 | ||
More Information
Set Membership: |
|

Dpy19l2
Genomic Map