Assay Details
| Gene Symbol: Dcps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153302.1 | NP_695214.1 | 3 - 4 | 517 | 100 | |
| GenBank mRNA | AF411249.1 | - | 3 - 4 | 517 | 100 | |
| BC089773.1 | - | 3 - 4 | 512 | 100 | ||
| BC105900.1 | - | 3 - 4 | 522 | 7377732 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dcps
Genomic Map