Assay Details
| Gene Symbol: Adamts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750455.3 | XP_003750503.1 | 6 - 7 | 2352 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts8| Gene Symbol: Adamts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750455.3 | XP_003750503.1 | 6 - 7 | 2352 | 110 | |
Set Membership: |
|