Assay Details
| Gene Symbol: Tirap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001055833.4 | XP_001055833.2 | 1 - 2 | 115 | 83 | |
| XM_006242794.2 | XP_006242856.1 | 1 - 2 | 115 | 83 | ||
More Information
Set Membership: |
|

Tirap
Genomic Map