Assay Details
| Gene Symbol: Rbm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053696.1 | NP_446148.1 | 1 - 2 | 83 | 85 | |
| XM_006256694.1 | XP_006256756.1 | 1 - 2 | 105 | 85 | ||
| XM_006256695.1 | XP_006256757.1 | 1 - 2 | 105 | 85 | ||
| XM_006256696.1 | XP_006256758.1 | 1 - 2 | 105 | 85 | ||
| GenBank mRNA | AF355190.1 | - | 1 - 2 | 47 | 85 | |
| BC087677.1 | - | 1 - 2 | 40 | 7318884 | 85 | |
| DY319481.1 | - | 1 - 2 | 83 | 85 | ||
| FQ220834.1 | - | 1 - 2 | 58 | 85 | ||
| FQ231739.1 | - | 1 - 2 | 58 | 85 | ||
| FQ235187.1 | - | 60 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF355190.1, BC087677.1, NM_053696.1, XM_006256694.1, XM_006256695.1, XM_006256696.1 transcripts.
Set Membership: |
|

Rbm3
Genomic Map