Assay Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191730.1 | NP_001178659.1 | 8 - 9 | 4196 | 58 | |
| XM_006256763.2 | XP_006256825.1 | 7 - 8 | 4273 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shroom4
Genomic Map