Assay Details
| Gene Symbol: Zfp184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751710.3 | XP_003751758.1 | 2 - 3 | 412 | 91 | |
| XM_006254113.2 | XP_006254175.1 | 2 - 3 | 479 | 91 | ||
| XM_006254114.2 | XP_006254176.1 | 2 - 3 | 604 | 91 | ||
| XM_008771768.1 | XP_008769990.1 | 2 - 3 | 604 | 91 | ||
| GenBank mRNA | BC128784.1 | - | 241 | 7381364 | 91 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC128784.1,XM_003751710.3,XM_006254113.2,XM_006254114.2,XM_008771768.1 transcripts.
Set Membership: |
|

Zfp184
Genomic Map