Assay Details
| Gene Symbol: Zfp184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751710.3 | XP_003751758.1 | 5 - 6 | 698 | 74 | |
| XM_006254113.2 | XP_006254175.1 | 5 - 6 | 765 | 74 | ||
| XM_006254114.2 | XP_006254176.1 | 5 - 6 | 890 | 74 | ||
| XM_006254118.2 | XP_006254180.1 | 2 - 3 | 621 | 74 | ||
More Information
Set Membership: |
|

Zfp184
Genomic Map