Assay Details
| Gene Symbol: Cabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108907.1 | NP_001102377.1 | 1 - 2 | 250 | 85 | |
| XM_006228328.2 | XP_006228390.1 | 1 - 2 | 268 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp5
Genomic Map