Assay Details
| Gene Symbol: Cabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108907.1 | NP_001102377.1 | 2 - 3 | 284 | 114 | |
| XM_006228328.2 | XP_006228390.1 | 2 - 3 | 302 | 114 | ||
| XM_006228329.2 | XP_006228391.1 | 2 - 3 | 602 | 114 | ||
| XM_006228330.2 | XP_006228392.1 | 2 - 3 | 507 | 114 | ||
| XM_006228331.2 | XP_006228393.1 | 2 - 3 | 1374 | 114 | ||
| XM_006228332.2 | XP_006228394.1 | 2 - 3 | 888 | 114 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228331.2,XM_006228332.2 transcripts.
Set Membership: |
|

Cabp5
Genomic Map