Assay Details
| Gene Symbol: LOC100911156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751978.3 | XP_003752026.1 | 2 - 3 | 297 | 127 | |
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109465.2 | NP_001102935.1 | 2 - 3 | 235 | 127 | |
| GenBank mRNA | AB983592.1 | - | 1 - 2 | 175 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911156
Genomic Map