Assay Details
| Gene Symbol: Abhd16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212531.2 | NP_997696.1 | 5 - 6 | 522 | 73 | |
| GenBank mRNA | BC091227.1 | - | 5 - 6 | 450 | 7317323 | 73 |
| CK653480.1 | - | 522 | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Abhd16a
Genomic Map